Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MYLK (Ser19) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 44260G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-260-G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-260-G Supplier Invitrogen™ Supplier No. 44260G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-MYLK (Ser19) Polyclonal Antibody for Western Blot

MYLK (MLCK) is a serine/threonine kinase that phosphorylates myosin regulatory light chains to assist in the production of contractile activity (myosin-actin interaction). MLCK is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MYLK (Ser19)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene MYLK
Gene Accession No. P11799, Q15746
Gene Alias 155 kda myosin light chain kinase homolog; 9530072E15Rik; A clone isolated from a chicken gizzard cDNA library which has high homology with the 3'-end sequence of the chicken gizzard myosin light chain kinase (MLCK) cDNA; The 5'-leader seqeuence derives from the intronic sequence of the MLCK gene.; A930019C19Rik; AAT7; AW489456; kinase related protein; kinase-related protein; KRP; KRP/Telokin; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; mylk; MYLK1; myosin light chain kinase; myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; nmMlck; smMLCK; smooth muscle myosin light chain kinase; smooth muscle myosin light chain kinase (61-kDa active fragment); smooth muscle myosin light chain kinase precursor (EC 2.7.2.37); telokin; unnamed protein product
Gene Symbols MYLK
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of MLC that contains serine 19 (serine 20 including the initiating methionine).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396445, 4638
Target Species Human, Chicken
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.