Learn More
Invitrogen™ Phospho-MYPT1 (Thr853) Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-MYPT1 (Thr853) |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | PPP1R12A |
| Gene Accession No. | O14974, Q10728, Q9DBR7 |
| Gene Alias | 1200015F06Rik; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase; 130 kDa myosin-binding subunit of smooth muscle myosin phophatase (M130); 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase; 130 kDa regulatory subunit of myosin phosphatase; 133kDa myosin-binding subunit of smooth muscle myosin phosphatase (M133); 5730577I22Rik; AA792106; AV099298; D10Ertd625e; hypothetical protein; I79_016312; LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 12A; M110; M130; M130 of smooth muscle myosin phosphatase; MBS; MBSP; myosin binding subunit; myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; myosin phosphatase-targeting subunit 1; myosin-binding subunit of myosin phosphatase; Mypt1; PP-1M; PP1M M110 subunit; PP1M subunit M110; Ppp1r12a; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1M 110 kda regulatory subunit; protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; si:dkey-28j4.1; smooth muscle myosin phosphatase myosin-binding subunit; unm p82emcf; unm_p82emcf; zgc:110448; zgc:110448 protein |
| Gene Symbols | PPP1R12A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.