Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Naked1 (Ser348) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105097 Supplier Invitrogen™ Supplier No. PA5105097
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of NAKED1 only when phosphorylated at Ser348.

In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Naked1 (Ser348)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NKD1
Gene Accession No. Q969G9, Q99MH6
Gene Alias 2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; naked-1; Nkd; NKD1; PP7246; Protein naked cuticle homolog 1
Gene Symbols NKD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NKD1(Accession Q969G9), corresponding to amino acid residues around phosphorylated Ser348.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364952, 85407, 93960
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.