Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-NFATC4 (Ser168, Ser170) Rabbit anti-Human∣Mouse, Polyclonal, Bioss
SDP

Catalog No. 501988627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8627 Supplier Bioss Supplier No. BS8535R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.

Specifications

Antigen Phospho-NFATC4 (Ser168, Ser170)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Nfatc4
Gene Accession No. Q14934, Q8K120
Gene Alias 3110041H08Rik; AW107667; AW546455; Nfat3; NF-AT3; Nfatc4; NF-ATc4; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4; nuclear factor of activated T-cells 4; Nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4
Gene Symbols Nfatc4
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human NFATC4 around the phosphorylation site of Ser168 + Ser170.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4776, 73181
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.