Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-p57 Kip2 (Thr310) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114613 Supplier Invitrogen™ Supplier No. PA5114613
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of p57 Kip2 only when phosphorylated at Thr310.

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-p57 Kip2 (Thr310)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CDKN1C
Gene Accession No. P49918
Gene Alias AL024410; Beckwith Wiedemann syndrome (WBS); BWCR; BWS; CDKI; Cdkn1c; cyclin dependent kinase inhibitor 1C; Cyclin dependent kinase inhibitor 1C (CDKN1C); cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (P57); cyclin-dependent kinase inhibitor 1C (p57, Kip2); cyclin-dependent kinase inhibitor p57; IMAGE; KIP2; p57; p57 Kip2; p57(kip2); p57KIP2; p57KIP2 A3sh; WBS
Gene Symbols CDKN1C
Host Species Rabbit
Immunogen A synthesized peptide derived from human p57 Kip2 around the phosphorylation site of Thr310.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.