Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p70 S6 Kinase Beta (Thr444, Ser447) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501985592
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5592 Supplier Bioss Supplier No. BS3700R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates the S6 ribosomal protein and eucaryotic translation initiation factor 4B. Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.

Specifications

Antigen Phospho-p70 S6 Kinase Beta (Thr444, Ser447)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rps6kb2
Gene Accession No. Q9UBS0, Q9Z1M4
Gene Alias 70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; Ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; Rps6kb2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; Serine/threonine-protein kinase 14B; SRK; STK14B
Gene Symbols Rps6kb2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human p70 S6 Kinase Beta around the phosphorylation site of Thr444/Ser447.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361696, 58988, 6199
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.