Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PAK4/6/7 (Ser474) Recombinant Rabbit Monoclonal Antibody (1H7L3)

Catalog No. PI702663 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI702663 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI702663 Supplier Invitrogen™ Supplier No. 702663
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Horse, Rat Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

PAK4 is highly expressed in prostate, testis and colon. PAK4 interacts tightly with GTP-bound but not GDP-bound CDC42 and weakly with RAC. PAK4 phosphorylates and autophosphorylates and also activates the JNK pathway. PAK5, a member of the PAK family of protein kinases contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK5 preferentially binds to CDC42 in the presence of GTP and the CRIB motif is essential for this interaction. PAK6 protein cotranslocates into the nucleus with androgen receptor, which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development, in response to androgen. PAK6 was initially identified as an androgen receptor in a yeast two hybrid screen and was found to be highly expressed in testis and prostate tissues. Later experiments have shown it to be activated by MAP kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAK4/6/7 (Ser474)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H7L3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Pak4
Gene Accession No. O96013, Q9NQU5, Q9P286
Gene Alias 2900083L08Rik; 4732456M09; 5730488L07Rik; 6430627N20; AW555722; KIAA1142; KIAA1264; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (CDKN1A)-activated kinase 6; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 4; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 6; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 4; p21(CDKN1A)-activated kinase 6; p21(CDKN1A)-activated kinase 7; p21-activated kinase 4; p21-activated kinase 5; p21-activated kinase 6; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 4/5/6; PAK 4/6/7; PAK4; PAK-4; PAK4/5/6; PAK5; PAK-5; PAK6; PAK-6; Pak7; PAK-7; protein kinase PAK5; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 6; serine/threonine-protein kinase PAK 6-like; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7
Gene Symbols Pak4, PAK5, PAK6
Host Species Rabbit
Immunogen Peptide corresponding to human Phospho-PAK4/PAK5/PAK6 (Ser474) (aa472-aa480).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10298, 56924, 57144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.