Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PAR1 (Tyr420) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105095 Supplier Invitrogen™ Supplier No. PA5105095
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Thrombin Receptor only when phosphorylated at Tyr420.

Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAR1 (Tyr420)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene F2R
Gene Accession No. P25116, P26824, P30558
Gene Alias AI482343; C HTR; Cf2r; coagulation factor II (thrombin) receptor; coagulation factor II receptor; coagulation factor II thrombin receptor; F2R; HTR; PAR1; PAR-1; protease activated receptor 1; protease-activated receptor 1; proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC
Gene Symbols F2R
Host Species Rabbit
Immunogen A synthesized peptide derived from human F2R(Accession P25116), corresponding to amino acid residues around phosphorylated Tyr420.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14062, 2149, 25439
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.