Learn More
Invitrogen™ Phospho-PCAF (Tyr729) Polyclonal Antibody

Description
Antibody detects endogenous levels of PCAF only when phosphorylated at Tyr729.
Specifications
Specifications
| Antigen | Phospho-PCAF (Tyr729) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KAT2B |
| Gene Accession No. | Q92831, Q9JHD1 |
| Gene Alias | A930006P13Rik; AI461839; AW536563; CAF; CREBBP-associated factor; histone acetylase PCAF; Histone acetyltransferase KAT2B; histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Kat2b; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Pcaf; Spermidine acetyltransferase KAT2B |
| Gene Symbols | KAT2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.