Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PDGFRA/PDGFRB (Tyr572, Tyr574) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 441000G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-100-0G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-100-0G Supplier Invitrogen™ Supplier No. 441000G
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Phospho-PDGFRA/PDGFRB (Tyr572, Tyr574) Polyclonal Antibody for Western Blot, ICC/IF

PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PDGFRA/PDGFRB (Tyr572, Tyr574)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1 mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene PDGFRB
Gene Accession No. P05622, P09619, P16234, P26618
Gene Alias AI115593; AI528809; alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; APDGFR; beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member A; CD140 antigen-like family member B; CD140a; CD140a antigen; CD140b; GAS9; IBGC4; IMF1; JTK12; KOGS; MGC74795; PDGF alpha chain; PDGF beta chain; PDGF Receptor alpha; PDGFACE; PDGFR; PDGFR1; PDGFR-1; PDGFR2; PDGFR-2; Pdgfra; pdgfr-a; PDGFRA/BCR fusion; PDGF-R-alpha; PDGFR-alpha; Pdgfrb; PDGF-R-beta; PDGFR-beta; PENTT; PGFRB; platelet derived growth factor receptor alpha; platelet derived growth factor receptor beta; platelet derived growth factor receptor, alpha polypeptide; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 1; platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; platelet-derived growth factor receptor, alpha polypeptide; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein; RHEPDGFRA
Gene Symbols PDGFRA, PDGFRB
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human PDGFRA that contains tyrosines 572 and 574 and PDGFRB tyrosines 579 and 581. The sequence is conserved in PDGFRA mouse, rat and frog.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18595, 18596, 5156, 5159
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.