Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PDGFRA/PDGFRB (Tyr849, Tyr857) Monoclonal Antibody (T.564.8)
GREENER_CHOICE

Catalog No. PIMA514905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514905 Supplier Invitrogen™ Supplier No. MA514905
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PDGFRA/PDGFRB (Tyr849, Tyr857)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone T.564.8
Concentration 632 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene PDGFRB
Gene Accession No. P05622, P09619, P16234, P20786, P26618, Q05030
Gene Alias AI115593; AI528809; alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; APDGFR; beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member A; CD140 antigen-like family member B; CD140a; CD140a antigen; CD140b; GAS9; IBGC4; IMF1; JTK12; KOGS; MGC74795; PDGF alpha chain; PDGF beta chain; PDGF Receptor alpha; PDGFACE; PDGFR; PDGFR1; PDGFR-1; PDGFR2; PDGFR-2; Pdgfra; pdgfr-a; PDGFRA/BCR fusion; PDGF-R-alpha; PDGFR-alpha; Pdgfrb; PDGF-R-beta; PDGFR-beta; PENTT; PGFRB; platelet derived growth factor receptor alpha; platelet derived growth factor receptor beta; platelet derived growth factor receptor, alpha polypeptide; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 1; platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; platelet-derived growth factor receptor, alpha polypeptide; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein; RHEPDGFRA
Gene Symbols PDGFRA, PDGFRB
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pTyr857 of human PDGF receptor β.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18595, 18596, 24629, 25267, 5156, 5159
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.