Learn More
Description
Recommended positive controls: Bovine cardiac preparation. PA1-25784 is expected to cross react with rabbit, mouse, rat, canine, and porcine due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-Phospholamban (Ser16) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 30% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PLN |
| Gene Accession No. | A4IFH6, P26678, P61015 |
| Gene Alias | cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product |
| Gene Symbols | PLN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
