Learn More
ABclonal Technology Phospho-PI3 Kinase p55-Y199
SDP
Supplier:  ABclonal Technology AP1463100UL
Encompass_Preferred
Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position The enzyme comprises a 110 kD catalytic subunit and a regulatory subunit of either 85 55 or 50 kD This gene encodes the 85 kD regulatory subunit Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin and a mutation in this gene has been associated with insulin resistance Alternative splicing of this gene results in four transcript variants encoding different isoforms
Catalog No. 50-000-14099
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.