Learn More
Invitrogen™ Phospho-PKC gamma (Thr514) Recombinant Rabbit Monoclonal Antibody (PKCgT514-PF4), FITC, Invitrogen™
Description
Specifications
Specifications
| Antigen | Phospho-PKC gamma (Thr514) |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | PKCgT514-PF4 |
| Conjugate | FITC |
| Formulation | PBS with 0.20% BSA and 0.09% sodium azide |
| Gene | PRKCG |
| Gene Accession No. | P05129, P63318, P63319 |
| Gene Alias | PKC; Pkcc; PKCG; PKCgamma; PKC-gamma; PKCI; Prkc; Prkcc; PRKCG; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma; RATPKCI; SCA14 |
| Gene Symbols | PRKCG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.