Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKC mu (Ser910) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114624 Supplier Thermo Scientific Supplier No. PA5114624
Only null left
Add to Cart
Add to Cart

Phospho-PKC mu (Ser910) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKC mu (Ser910)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Prkd3
Gene Accession No. O94806, Q15139, Q62101, Q8BZ03, Q8K1Y2, Q9BZL6
Gene Alias 4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3
Gene Symbols PRKD1, Prkd2, Prkd3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKD1/PKC mu around the phosphorylation site of Ser910.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101540, 18760, 23683, 25865, 5587, 75292
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.