Learn More
Invitrogen™ Phospho-PRAS40 (Thr356) Polyclonal Antibody

Description
Antigen Affinity Purified from Pooled Serum.
Specifications
Specifications
| Antigen | Phospho-PRAS40 (Thr356) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | PRAS40 |
| Gene Accession No. | Q7JXA2 |
| Gene Alias | 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 |
| Gene Symbols | PRAS40 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.