Learn More
Invitrogen™ Phospho-Rb (Ser780) Recombinant Rabbit Monoclonal Antibody (2E9)

Description
Phospho-Rb (Ser780) Recombinant Monoclonal Antibody for ELISA, ICC/IF, IHC (P), IP
Specifications
Specifications
| Antigen | Phospho-Rb (Ser780) |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 2E9 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05 BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RB1 |
| Gene Accession No. | P06400 |
| Gene Alias | Cleaved Rb; delta RB-p70; DRb-p70; exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; OSRC; p104; p104 chicken Rb; P105RB; p105-Rb; Phospho-Retinoblastoma; pp105; pp110; PPP1R130; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; rb; RB transcriptional corepressor 1; RB1; Rb-1; Rb-p70; Retin; Retinoblastoma; retinoblastoma 1; retinoblastoma 1 (including osteosarcoma); retinoblastoma 1 protein; retinoblastoma gene; retinoblastoma isoform A; retinoblastoma protein; retinoblastoma suspectibility protein; retinoblastoma tumor suppressor; retinoblastoma-associated protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.