Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RRN3 (Ser649) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103273 Supplier Invitrogen™ Supplier No. PA5103273
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TIF1 alpha only when phosphorylated at Ser649.

Required for efficient transcription initiation by RNA polymerase I. Required for the formation of the competent preinitiation complex (PIC). Dissociates from pol I as a consequence of transcription. In vitro, cannot activate transcription in a subsequent transcription reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RRN3 (Ser649)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RRN3
Gene Accession No. B2RS91, Q9NYV6
Gene Alias A-270G1.2; AL023001; E130302O19Rik; R75565; RGD1305001; RNA polymerase I-specific transcription initiation factor RRN3; Rrn3; RRN3 homolog, RNA polymerase I transcription factor; RRN3 RNA polymerase I transcription factor homolog; RRN3 RNA polymerase I transcription factor homolog (S. cerevisiae); RRN3 RNA polymerase I transcription factor homolog (yeast); Tif1a; TIF-1A; TIFIA; TIF-IA; Transcription initiation factor IA; transcription initiation factor TIF-IA
Gene Symbols RRN3
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRN3(Accession Q9NYV6), corresponding to amino acid residues around phosphorylated Ser649.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106298, 304714, 54700
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.