Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-S6 (Ser235, Ser236) Monoclonal Antibody (cupk43k), eFluor™ 450, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501124219
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-4219 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-4219 Supplier Invitrogen™ Supplier No. 48900742
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This cupk43k monoclonal antibody recognizes human and mouse ribosomal protein S6 (also known as 40S ribosomal protein S6, phosphoprotein NP33, RPS6, RS6, S6) when phosphorylated on serine 235 (S235, human) and serine 236 (S236, mouse). Ribosomal protein S6 is a component of the 40S subunit of the ribosome and is phosphorylated at multiple sites following stimulation of cells by growth factors, tumor promoting agents, or mitogens. Phosphorylation of ribosomal protein S6 by p70S6K and PKDCD results in upregulation of the translation of RNA coding for proteins involved in cell cycle entry. Ribosomal protein S6 is dephosphorylated upon growth arrest. The specificity of the cupk43k monoclonal antibody was determined by western blotting. Applications Reported: This cupk43k antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This cupk43k antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-S6 (Ser235, Ser236)
Applications Flow Cytometry
Classification Monoclonal
Clone cupk43k
Concentration 5 μL/Test
Conjugate eFluor 450
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene RPS6
Gene Accession No. P62753, P62754
Gene Alias (Rp)S6; 40S ribosomal protein S6; air[8]; air8; anon-WO02059370.61; CG10944; CG10944-PA; CG10944-PB; Dmel\CG10944; Dmel_CG10944; DS6; hemocytes enlarged; hen; l(1)air[8]; l(1)air8; l(1)hen; LD31286p; lethal(1)aberrant immune response[8]; lethal-(1)-haemocytes enlarged; lethal-aberrant-immune-response-8; M(1)7BC; M(1)7C; minute; minute (1) 7BC; OK/SW-cl.2; OTTHUMP00000021121; OTTHUMP00000021122; Phosphoprotein NP33; pp30; ribosomal protein S6; Rp S6; RP11-513M16.6; RPS6; RpS6-PA; RpS6-PB; RS6; S6; S6 ribosomal protein; S6R; Small ribosomal subunit protein eS6; wu:fa92e06; wu:fb64g06; zgc:92237
Gene Symbols RPS6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20104, 6194
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

How does eBioscience determine the specificity of their phosphorylation-specific antibodies?

Antibodies are tested and validated in a variety of ways. First, we verify that in a panel of cells in which different pathways have been induced, we see phosphorylation specific staining only in the cells in which the specific pathway of interest has been activated. Second, we verify that phosphorylation specific staining is observed only in cell types in which the protein is expressed and not in cell types in which the protein is not expressed. Third, whenever possible, western immunoblotting is used to confirm the presence of a band(s) of the appropriate size(s) in stimulated/treated cells (and not in unstimulated/untreated cells, as appropriate).

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

How long can cells that have been fixed and placed in methanol be stored for use with eBioscience antibodies? What is the recommended storage temperature? How about storage of fixed cells in eBioscience fixation buffers?

We have data demonstrating that fixed cells stored in methanol are stable at -20 degrees C or at -80 degrees C for several weeks. We do not recommend storing fixed cells in eBioscience Foxp3/Transcription Factor Buffer. However, cells fixed with eBioscience Foxp3/Transcription Factor Buffer or eBioscience Intracellular (IC) fixation buffer can be stored in eBioscience IC fixation buffer for up to 3 days at 4 degrees C in the dark.
Note: Please be aware that higher compensation values may be seen with tandem dyes if any other fixative is used apart from eBioscience IC fixation buffer, for 3-day storage.

What is the optimal protocol for eBioscience phosphorylation-specific antibodies?

Each antibody has been tested in three different buffer systems: IC fixation and permeabilization, Foxp3/Transcription Factor Buffer, and IC fixation Buffer/Methanol. The recommended buffer system(s) will be noted on the Technical Data Sheet for the specific antibody.

What is the best way to maintain surface staining when using the methanol-based protocol?

Because methanol can destroy the epitope recognized by some antibodies, ideally surface staining would be performed at the beginning of the protocol with a methanol-resistant fluorochrome (like eFluor 450, FITC, and eFluor 660).
Please recognize that surface staining before stimulation can have undesired effects due to activation of the cell caused by antibody binding. We recommend staining after stimulation/treatment, fixation, and methanol treatment.

Can I use other buffer systems from other companies when using eBioscience phosphorylation-specific antibodies?

In general, you can use other companies' buffer systems provided that their buffers are of a similar composition as the buffers recommended. Be aware that results will vary depending upon the buffer system used. eBioscience antibodies have been optimized for use in eBioscience buffers and we have not tested all of our antibodies in other buffer systems. Thus, we highly recommend using our optimized protocol and buffer systems.

What are the appropriate controls to run when using eBioscience phosphorylation-specific antibodies?

It is important to understand whether the stimulation/treatment results in an upregulation or a down regulation of the phosphorylation event. Thus, it is recommended that changes in phosphorylation levels in stimulated/treated versus unstimulated/untreated cells be compared. A histogram or density plot overlay these two treatments will provide a better assessment of a change in phosphorylation event than using isotype controls.

Can I perform phosphorylation specific staining and stain for intracellular cytokines at the same time when using eBioscience phosphorylation-specific antibodies?

In theory, if the buffer systems are compatible for the antibodies of interest, this can be done. In practice, cytokine translation and phosphorylation events do not typically occur in the same timeframe, therefore, it may be difficult to detect both events in the same sample. The results will be depended on the kinetics and should be determined for each system and protein of interest.

When using eBioscience phosphorylation-specific antibodies, can I perform phosphorylation specific staining together with transcription factor staining at the same time? If so, what buffer and protocol do I use?

In theory, this can be done if the antibodies against both the transcription factor and the phosphorylated protein work in the same buffer system. For example, Anti-Human/Mouse phsopho-H2AX and all transcription factor eBioscience antibodies offered will work in the Foxp3/Transcription Factor Buffer System (Cat. No. 00-5523-00).

For eBioscience phosphorylation-specific antibodies, can total and phosphorylated protein be analyzed in parallel?

In theory, this can be done if the antibodies to both proteins work in the same buffer system and if the antibodies recognize different epitopes. Please refer to the individual Technical Data Sheets for information about buffer compatibility. Also, consider using eBioscience InstantOne ELISA Kits for immunoassay quantitation of both total and phosphorylated proteins such as AKT1/2/3, JNK1/2/3, NFκB, STAT3, and STAT5.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.