Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SAR1A (Thr139) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105081 Supplier Invitrogen™ Supplier No. PA5105081
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of SAR1A only when phosphorylated at Thr139.

Involved in transport from the endoplasmic reticulum to the Golgi apparatus. Required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SAR1A (Thr139)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SAR1A
Gene Accession No. P36536, Q9NR31
Gene Alias 1600019H17Rik; COPII-associated small GTPase; GTP-binding protein SAR1a; masra2; SAR1; SAR1 gene homolog A; SAR1 gene homolog A (S. cerevisiae); SAR1 homolog A; SAR1A; SAR1a gene homolog 1; Sara; Sara1; secretion associated Ras related GTPase 1A; secretion associated, Ras related GTPase 1A
Gene Symbols SAR1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human SAR1A(Accession Q9NR31), corresponding to amino acid residues around phosphorylated Thr139.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20224, 361842, 56681
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.