Learn More
Invitrogen™ Phospho-(Ser/Thr) ATM + ATR Substrate Monoclonal Antibody (G.770.10)

Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with corresponding nonphosphorylated sequences or with other phospho-Ser/Thr-containing motifs. This antibody preferentially binds peptides and proteins that contain phospho-Ser/Thr preceded by Leu or similar hydrophobic amino acids at the -1 position and followed by Gln at the +1 position.
Specifications
Specifications
| Antigen | Phospho-(Ser/Thr) ATM + ATR Substrate |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | G.770.10 |
| Concentration | 192 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene Alias | L(s/t)Q; SQ/TQ |
| Host Species | Rabbit |
| Immunogen | Synthetic phospho-ATM/ATR substrate peptides. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.