Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SGK1 (Ser422) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 441264G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-126-4G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-126-4G Supplier Invitrogen™ Supplier No. 441264G
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Phospho-SGK1 (Ser422) Polyclonal Antibody for Western Blot, IHC (P)

SGK (SGK1) is a serine/threonine kinase that may be a transcriptional target for p38/MAPK in hepatoma cells. SGK plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SGK1 (Ser422)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene SGK1
Gene Accession No. O00141
Gene Alias serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3
Gene Symbols SGK1
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human SGK1 that contains serine 422.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6446
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.