Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SMAD2/SMAD3 (Ser465, Ser467, Ser423, Ser425) Recombinant Rabbit Monoclonal Antibody (ST0493)

Catalog No. MA556386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556386 Supplier Invitrogen™ Supplier No. MA556386
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Positive Control: A549 treated with 5 ng/mL TGF-beta1 for 24 hours whole cell lysate, C2C12 treated with 5 ng/mL TGF-beta1 for 24 hours whole cell lysate, HeLa starved overnight then treated with 10 ng/mL TGF-beta1 for 30 minutes cell lysate, NIH/3T3 starved overnight then treated with 10 ng/mL TGF-beta1 for 30 minutes cell lysate, A549, human large intestine tissue, mouse skin tissue, mouse kidney tissue. Tissue Specificity: Low tissue specificity. Subcellular Location: Cytoplasm, Nucleus. Sequence Similarities: 100% Mouse. Predicted band size: 48 kDa.

Smad proteins, the mammalian homologs of the Drosophila mothers against decapentaplegic (Mad), have been implicated as downstream effectors of TGFβ/BMP signaling. Smad1 (also designated Madr1 or JV4-1) and Smad5 are effectors of BMP-2 and BMP-4 function, while Smad2 (also designated Madr2 or JV18-1) and Smad3 are involved in TGFβ and Activin-mediated growth modulation. Smad4 (also designated DPC4) has been shown to mediate all of the above activities through interaction with various Smad family members. Smad6 and Smad7 regulate the response to Activin/TGFβ signaling by interfering with TGFβ-mediated phosphorylation of other Smad proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SMAD2/SMAD3 (Ser465, Ser467, Ser423, Ser425)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone ST0493
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SMAD2
Gene Accession No. P84022, Q15796, Q62432, Q8BUN5
Gene Alias 7120426M23Rik; AU022421; hMAD-2; hMAD-3; hSMAD2; hSMAD3; HSPC193; HsT17436; JV15-2; JV18; JV18-1; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 2; MAD homolog 3; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MADH2; MADH3; MADR2; mad-related protein 2; mMad2; mMad3; mother against DPP homolog 2; mothers against decapentaplegic homolog 2; Mothers against decapentaplegic homolog 3; mothers against DPP homolog 2; mothers against DPP homolog 3; pSMAD2; Sma- and Mad-related protein 2; SMA- and MAD-related protein 3; SMAD; Smad 2; Smad 3; SMAD family member 2; SMAD family member 3; SMAD, mothers against DPP homolog 2; SMAD, mothers against DPP homolog 3; Smad2; Smad-2; Smad3
Gene Symbols SMAD2, SMAD3
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser423 and 425 of human Smad3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17126, 17127, 4087, 4088
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.