Learn More
Invitrogen™ Phospho-STAT1 (Tyr701) Monoclonal Antibody (S.213.5)
Shop All Thermo Scientific Products

Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the corresponding phospho-tyrosines of other Stat proteins.
Specifications
Specifications
| Antigen | Phospho-STAT1 (Tyr701) |
| Applications | ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | S.213.5 |
| Concentration | 59 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 |
| Gene | STAT1 |
| Gene Accession No. | P42224, P42225 |
| Gene Alias | 2010005J02Rik; AA408197; CANDF7; DD6G4-4; DKFZp686B04100; IMD31A; IMD31B; IMD31C; ISGF-3; LOW QUALITY PROTEIN: signal transducer and activator of transcription 1-alpha/beta; OTTHUMP00000165046; OTTHUMP00000205845; signal transducer and activator of transcription 1; signal transducer and activator of transcription 1, 91kD; signal transducer and activator of transcription 1, 91kDa; signal transducer and activator of transcription 1-alpha/beta; signal transducer and activator of transcription-1; STAT; STAT 1; STAT1; STAT-1; stat1 alpha; STAT91; transcription factor ISGF-3; transcription factor ISGF-3 components p91/p84 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.