Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Stat5 (Tyr694) Recombinant Rabbit Monoclonal Antibody (Stat5Y694-G11), Invitrogen™

Catalog No. p-7215092
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537354 20 μL
PIMA537108 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537354 Supplier Invitrogen™ Supplier No. MA537354
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Stat5 (Tyr694)
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone Stat5Y694-G11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
Gene STAT5A
Gene Accession No. P42229, P42230
Gene Alias AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5
Gene Symbols STAT5A
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr694 of human phospho Stat5.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20850, 6776
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.