Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human STAT5 alpha/beta (Phospho) [pY694/pY699] InstantOne™ ELISA Kit

Catalog No. 50246264
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-264 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-246-264 Supplier Invitrogen™ Supplier No. 858611211
Add to Cart
Edge
Add to Cart

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human STAT5A/B in cell lysates. Detection of mouse and rat STAT5A/B is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor-associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of, many cell ligands, such as IL-2, IL-3, IL-7, GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/ BCL-X(L), which suggests the antiapoptotic function of this gene in cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42229,P51692
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6776, 6777
Gene Symbol STAT5,MGF,STAT5
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Synonym Signal Transducer and Activator of Transcription 5
Target STAT5 alpha/beta
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 10 mins
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.