Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Synaptotagmin 1 (Ser309) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537750 Supplier Invitrogen™ Supplier No. PA537750
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for ICC/IF is Hela cells.

Synaptotagmin 1 is a synaptic vesicle membrane glycoprotein that is widely expressed throughout the CNS and is generally thought to act as the Ca2+ sensor in the regulation of exocytosis and neurotransmitter release. Studies indicate that synaptotagmin's Ca2+ mediated binding of SNAP25 is essential for the Ca2+ dependent triggering of membrane. It has been demonstrated that discrete residues within the c(2)b binding domain of synaptotagmin 1 independently specify endocytic rate and synaptic vesicle size. Synaptotagmin 1 may have a regulatory role in the membrane interactions during trafficking of synaptic vesicles at the active zone of the synapse. Further, Synaptotagmin 1 binds acidic phospholipids with a specificity that requires the presence of both an acidic head group and a diacyl backbone. A Ca(2+)-dependent interaction between synaptotagmin and putative receptors for activated protein kinase C has been reported. Synaptotagmin 1 can also bind to at least three additional proteins in a Ca(2+)-independent manner; these are neurexins, syntaxin and AP2. Diseases associated with the dysfunction of Synaptotagmin 1 include food-borne botulism and infant botulism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Synaptotagmin 1 (Ser309)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Syt1
Gene Accession No. P21579, P21707, P46096
Gene Alias AW124717; DKFZp781D2042; FLJ42519; G630098F17Rik; P65; SV DKFZp781D2042; SV SYT; svp65; Syb1; synaptotagmin 1; synaptotagmin 1 L homeolog; synaptotagmin 2; synaptotagmin I; synaptotagmin I protein; synaptotagmin I variant b; Synaptotagmin II; Synaptotagmin XI; synaptotagmin-1; Synaptotagmin-2; SYT; Syt1; syt1.L; SYT2; SytI; SytII; XELAEV_18022019mg
Gene Symbols Syt1
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of serine 309 (G-L-S(p)-D-P) derived from Human Synaptotagmin 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20979, 25716, 6857
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.