Learn More
Invitrogen™ Phospho-TERT (Ser227) Polyclonal Antibody

Description
PA5-38817 has been used successfully in peptide-ELISA at a dilution of 1:1,000.
Specifications
Specifications
| Antigen | Phospho-TERT (Ser227) |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TERT |
| Gene Accession No. | O14746 |
| Gene Alias | CMM9; DKCA2; DKCB4; EST2; HEST2; hTERT; hTRT; PFBMFT1; RGD:70494}; TCS1; Telomerase catalytic subunit; telomerase reverse transcriptase; telomerase reverse transcriptase beta; telomerase-associated protein 2; TERT; tert {ECO:0000312; TP2; TR; TRT |
| Gene Symbols | TERT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.