Learn More
Invitrogen™ Phospho-TIRAP (Tyr86) Polyclonal Antibody

Description
This antibody is predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-TIRAP (Tyr86) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | TIRAP |
| Gene Accession No. | P58753, Q99JY1 |
| Gene Alias | AA407980; adapter protein wyatt; adaptor protein Wyatt; BACTS1; C130027E04Rik; FLJ42305; HGNC:17192; Mal; myD88 adapter-like protein; MyD88-2; OTTHUMP00000179130; TIR domain containing adaptor protein; TIR domain-containing adapter protein; TIRAP; Tlr4ap; toll/interleukin-1 receptor domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; toll-interleukin 1 receptor (TIR) domain-containing adaptor protein; Toll-like receptor 4 adaptor protein; Toll-like receptor adaptor protein; Wyatt |
| Gene Symbols | TIRAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.