Learn More
Invitrogen™ Phospho-TPL2 (Thr290) Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 53 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | Phospho-TPL2 (Thr290) |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MAP3K8 |
| Gene Accession No. | P41279, Q07174, Q63562 |
| Gene Alias | augmented in rheumatoid arthritis 2; AURA2; Cancer Osaka thyroid oncogene; cancer Osaka thyroid, oncogene; CCOT; c-COT; COT; cot (cancer Osaka thyroid) oncogene; COT proto-oncogene serine/threonine-protein kinase; Cot/Tpl2; D17TPL2; EC 1.14.16.2; Est; Estf; Ewing sarcoma transformant; FLJ10486; M3K8; MAP3K 8; Map3k8; MEKK8; mitogen activated protein kinase kinase kinase 8; mitogen-activated protein kinase kinase kinase 8; Protein kinase cot; Proto-oncogene c-Cot; proto-oncogene serine/threoine protein kinase; serine/threonine kinase (Tpl-2); serine/threonine-protein kinase cot; TH isoform 3; TH isoform a; TH-4; TPL 2; Tpl2; TPL-2; Tumor progression locus 2; TY3H; TYH |
| Gene Symbols | MAP3K8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.