Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TRAP220 (Thr1032) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106026 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106026 Supplier Invitrogen™ Supplier No. PA5106026
  • $587.00
    $489.65 / Each of 1

    Save $97.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TRAP220/MED1 only when phosphorylated at Thr1032.

Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TRAP220 (Thr1032)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MED1
Gene Accession No. Q15648, Q925J9
Gene Alias Activator-recruited cofactor 205 kDa component; AI480703; ARC205; CRSP1; CRSP200; CRSP210; DRIP205; DRIP230; l11Jus15; MED1; Mediator complex subunit 1; mediator of RNA polymerase II transcription subunit 1; p53 regulatory protein RB18A; Pbp; peroxisome proliferator activated receptor binding protein; peroxisome proliferator-activated receptor-binding protein; Ppar binding protein; PPAR-binding protein; PPARBP; PPARG binding protein; PPARGBP; PPRB; RB18A; RGD1559552; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; thyroid receptor-interacting protein 2; TRAP220; TR-interacting protein 2; TRIP2; TRIP-2; vitamin D receptor-interacting protein 230 kD; vitamin D receptor-interacting protein complex component DRIP205
Gene Symbols MED1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MED1(Accession Q15648), corresponding to amino acid residues around phosphorylated Thr1032.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19014, 5469
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.