Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516) Rabbit anti-Human∣Mouse, Polyclonal, Bioss
SDP

Catalog No. 501986626
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6626 Supplier Bioss Supplier No. BS5525R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen Phospho-TrkA/B/C (Tyr496, Tyr516, Tyr516)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NTRK1
Gene Accession No. P04629, P15209, Q16620, Q3UFB7
Gene Alias BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C80751; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; High affinity nerve growth factor receptor; MTC; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 1; neurotrophic receptor tyrosine kinase 2; Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; NTRK1; NTRK2; Oncogene TRK; p140-TrkA; RATTRKB1; Slow nerve growth factor receptor; Tkr; Tkrb; TRK; TRK1; TRK1-transforming tyrosine kinase protein; TRKA; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TRKB; Trk-B; TrkB tyrosine kinase; TRKB1; Tropomyosin-related kinase A; Tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B
Gene Symbols NTRK1, Ntrk2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from rat NTRK2 around the phosphorylation site of Tyr516.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18211, 18212, 4914, 4915
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.