Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TYK2 (Tyr1054) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105746 Supplier Invitrogen™ Supplier No. PA5105746
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TYK2 only when phosphorylated at Tyr1054.

Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TYK2 (Tyr1054)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TYK2
Gene Accession No. P29597, Q9R117
Gene Alias 4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; IMD35; JTK1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; Tyk2; tyrosine kinase 2; tyrosine kinase TYK2
Gene Symbols TYK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TYK2(Accession P29597), corresponding to amino acid residues around phosphorylated Tyr1054.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54721, 7297
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.