Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-VAV2 (Tyr172) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143788 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143788 Supplier Invitrogen™ Supplier No. PA5143788
  • $579.00
    $537.65 / Each of 1

    Save $41.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated Vav2 (Tyr-172) peptide before affinity purification using phospho-Vav2 (Tyr-172) peptide (without carrier). The antibody detects a 100 kDa* band corresponding to Vav on SDS-PAGE immunoblots of human A431 cells treated with EGF and detects Vav1, Vav2, and Vav3 in Jurkat cells treated with pervanadate. The antibody does not detect these Vavs after treatment with alkaline phosphatase.

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-VAV2 (Tyr172)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.125 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Vav2
Gene Accession No. P52735, Q60992
Gene Alias 2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene
Gene Symbols Vav2
Host Species Rabbit
Immunogen Phospho-Vav2 (Tyr-172) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Tyr-172 in mouse Vav2. This sequence has high homology with similar regions in rat and human Vav2, and has significant homology to the conserved site in Vav1 and Vav3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22325, 296603, 7410
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.