Learn More
Invitrogen™ Phospho-Vimentin (Ser39) Recombinant Rabbit Monoclonal Antibody (3H9L2)
Shop All Thermo Scientific ProductsDescription
This antibody is predicted to react with Monkey, Cat, Pig and Sheep Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | Phospho-Vimentin (Ser39) |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3H9L2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | BRIP1 |
| Gene Accession No. | Q9BX63 |
| Gene Alias | 3110009N10Rik; 8030460J03Rik; ATP-dependent RNA helicase BRIP1; BACH 1; Bach1; BACH1 BRIP1; BRCA1 interacting protein C-terminal helicase 1; BRCA1/BRCA2-associated helicase 1; BRCA1-associated C-terminal helicase 1; BRCA1-binding helicase-like protein BACH1; BRCA1-interacting protein 1; BRCA1-interacting protein C-terminal helicase 1; BRIP 1; Brip1; FACJ; FANCJ; FANCJ protein; Fanconi anemia group J protein; Fanconi anemia group J protein homolog; FLJ90232; MGC126521; MGC126523; OF; Protein FACJ |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.