Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Wee1 (Ser53) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986667
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6667 Supplier Bioss Supplier No. BS5589R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Wee1 is a nuclear protein, which is a tyrosine kinase belonging to the Ser/Thr family of protein kinases. This protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.

Specifications

Antigen Phospho-Wee1 (Ser53)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene WEE1
Gene Accession No. P30291
Gene Alias BOS_14491; DKFZp686I18166; fa20h03; FLJ16446; wee 1 homolog; WEE 1 homolog 1 (S. pombe); WEE1; WEE1 G2 checkpoint kinase; WEE1 homolog; WEE1 homolog (S. pombe); wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; Wee1A kinase; WEE1hu; Wee1-like protein kinase; wu:fa20h03; zgc:136514
Gene Symbols WEE1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Wee1 around the phosphorylation site of Ser53 [ED (p-S)AF].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7465
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.