Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-WIPI2 (Ser413) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104678 Supplier Invitrogen™ Supplier No. PA5104678
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of WIPI2 only when phosphorylated at Ser413.

WD repeat proteins play a role in many essential biologic functions, regulating the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. WIPI2, also known as ATG18B or ATG21, is a human homolog to yeast ATG18 and contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. It is recruited to early autophagosomal structures along with Atg16L and ULK1 and is required for the formation of LC3-positive autophagosomes. Along with the highly related WIPI1, WIPI2 is found at the plasma membrane in addition to autophagosomal membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-WIPI2 (Ser413)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WIPI2
Gene Accession No. Q6AY57, Q80W47, Q9Y4P8
Gene Alias 1110018O08Rik; 2510001I10Rik; ATG18B; Atg21; CGI-50; WD repeat domain phosphoinositide-interacting protein 2; WD repeat domain, phosphoinositide interacting 2; WD40 repeat protein interacting with phosphoinositides 2; WIPI2; WIPI-2; WIPI49-like protein 2
Gene Symbols WIPI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human WIPI2(Accession Q9Y4P8), corresponding to amino acid residues around phosphorylated Ser413.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26100, 288498, 74781
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.