Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phosphodiesterase II (Bovine)

Catalog No. 89956748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-748 10 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-748 Supplier Abnova™ Supplier No. P5253
Add to Cart
Add to Cart

Used for Enzyme

Specifications

For Use With (Application) Enzyme Activity
Name Phosphodiesterase II (Bovine)
Preparation Method Native protein purified from Bovine Spleen
Quantity 10 Units
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name PHOSPHODIESTERASE
Assays Phosphodiesterase II Assay
Biological Activity ≥1.2μ/mg dry weight. One Unit increases the absorbance at 260nm by 0.200 in 30 minutes at 37°C, pH 6.5, with an RNA substrate.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Spleen
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.