Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic acid, trisodium salt hydrate, MP Biomedicals™

Catalog No. ICN15187290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15187290 500 mg
ICN15187225 25 mg
ICN15187280 100 mg
ICN15187283 250 mg
ICN15187291 1 g
ICN15187294 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN15187290 Supplier MP Biomedicals, Inc Supplier No. 0215187290
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5541-93-5
Molecular Formula C3H2Na3O6P
Molecular Weight (g/mol) 233.986
InChI Key RJUFBDMHZGRGMA-UHFFFAOYSA-K
Synonym 2-propenoic acid, 2-phosphonooxy-, trisodium salt, trisodium 2-phosphonatooxy prop-2-enoate, unii-78y3q07j1x, phosphoenolpyruvate trisodium salt, 2-phosphoenol pyruvate na3salt, 2-propenoic acid, 2-phosphonooxy-, sodium salt 1:3, trisodium phosphonatoenolpyruvate, trisodium 2-phosphonatoacrylate, trisodium phosphoenolpyruvate, c3h2o6p.3na
PubChem CID 79673
IUPAC Name trisodium;2-phosphonatooxyprop-2-enoate
SMILES C=C(C(=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Quantity 500 mg
Formula Weight 252
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic acid, trisodium salt hydrate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.