Learn More
Bioassay Systems EnzyChrom™ Phospholipid Assay Kit. For quantitative determination of phospholipid and evaluation of drug effects on its metabolism.
SDP
Catalog No. 50489287 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of phospholipid and evaluation of drug effects on its metabolism. Key Features: Sensitive. Use 20 µL samples. Linear detection range: colorimetric assay 3 - 200 µM, fluorimetric assay 0.6 - 20 µM phospholipid. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: OD, FL: 3, 0.6 µM.
Catalog No. 50-489-287 Supplier Bioassay Systems Supplier No. EPLP100
For quantitative determination of phospholipid and evaluation of drug effects on its metabolism. Key Features: Sensitive. Use 20 µL samples. Linear detection range: colorimetric assay 3 - 200 µM, fluorimetric assay 0.6 - 20 µM phospholipid. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: OD, FL: 3, 0.6 µM.