Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 10% (v/v), Ricca Chemical
SDP

Catalog No. 585055 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
55 gal.
Packaging:
Poly Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
585055 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 585055 Supplier Ricca Chemical Company Supplier No. 585055
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
Concentration 10% (v/v)
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Poly Drum
Solvent or Matrix Aqueous Solution
Quantity 55 gal.
Physical Form Liquid
Percent Purity 10% V/V
Grade Laboratory
Chemical Name or Material Phosphoric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.