Learn More
LAB ALLEY Lab Alley Phosphoric Acid 25% Solution Food Grade (FCC) Kosher Halal
SDP
Supplier:  LAB ALLEY PHAF2555GAL
Encompass
Phosphoric acid also known as orthophosphoric acid is a weak triprotic acid. At 25% it is a colorless and odorless liquid. The FCC Food Grade designation ensures that this product complies with the purity standards set by the United States Pharmacopeia and is therefore suitable for food use. Kosher certification requires that all ingredients equipment and processing comply with the Jewish dietary law.
Catalog No. NC2629471