Learn More
Medchemexpress LLC Phosphoric acid, 4-(1-methylethyl)phenyl diphenyl ester | 55864-04-5 | MFCD00377267 | 95.0% | 368.36 g/mol | C21H21O4P | 25 MG

Supplier: Medchemexpress LLC HYW58826825MG
4-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant used as an additive in materials research and flame-retardant formulations. It is identified by CAS 55864-04-5, has molecular formula C21H21O4P, and a molecular weight of 368.36 g/mol. The substance is supplied as a research reagent for laboratory use.
- Organophosphate flame retardant additive for materials research.
- Molecular formula C21H21O4P; molecular weight 368.36 g/mol.
- CAS number 55864-04-5 for unambiguous identification.
- Available in small research quantities such as 25 mg.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.