Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 4M in 1-Propanol, Reagents

Catalog No. 01334127
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-127 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-127 Supplier Reagents Holdings Llc Supplier No. CP2300004C
Add to Cart
Add to Cart

Phosphoric Acid, 4M in 1-Propanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-23-8 , 7664-38-2
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Color Colorless to Amber
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.0452
Quantity 4 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Phosphoric Acid, 4M in 1-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.