Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 50% (v/v), Ricca Chemical
SDP

Catalog No. R585300010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R585300010F 10 L Cubitainer
58531 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R585300010F Supplier Ricca Chemical Company Supplier No. 58532.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Cubitainer
Assay Percent Range 50.0 ± 0.5% v/v
Solvent or Matrix Aqueous Solution
Quantity 10 L
Percent Purity 50% V/V
Grade Laboratory
Chemical Name or Material Phosphoric Acid, 50% v/v (1+1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.