Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RABP00301C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Amber Glass Bottle
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABP00301C 1 L Amber Glass Bottle
RABP0030500 500 mL Glass Bottle
RABP003025D 2.5 L Glass Bottle
RABP00304C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RABP00301C Supplier Ricca Chemical Company Supplier No. RABP00301C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid

Specifications

Color Colorless
CAS Min % 85.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging Amber Glass Bottle
Reducing Substances Pass
Quantity 1 L
Synonym orthophosphoric acid
SMILES OP(O)(O)=O
Molecular Weight (g/mol) 97.99
Percent Purity ≥85%
Grade ACS
Chemical Name or Material Phosphoric Acid
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.