Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), Reagents

Catalog No. 01336818
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-818 500 mL Glass Bottle
01-336-824 1 L Poly Bottle
01-336-823 2.5 L Poly Bottle
01-336-822 2.5 L Glass Bottle
01-336-820 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-336-818 Supplier Reagents Holdings Llc Supplier No. C5101950500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 260°C
Packaging Glass Bottle
Specific Gravity 1.69
Quantity 500 mL
Physical Form Liquid
Grade ACS
Chemical Name or Material Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.