Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric acid, Solstice

Catalog No. 60090462 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-462 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-462 Supplier Solstice Supplier No. 304171LGL
Add to Cart
Add to Cart

Puriss. p.a., ACS reagent, Reag. ISO, Reag. Ph. Eur., ≥85%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
PubChem CID 1004
ChEBI CHEBI:26078

Specifications

Packaging Glass Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity ≥85 %
Grade ACS
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.