Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, NF, 85-88%, Spectrum™ Chemical
SDP

Catalog No. 18605178 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-178 500 mL Amber Glass Bottle
18-605-182 2.5 L Glass Bottle
18-605-181 2.5 L Poly-Coated Glass Bottle
18-605-179 6 x 500 mL Amber Glass Bottle
18-605-183 4 L Poly-Coated Glass Bottle
18-605-184 6 x 1 L Amber Glass Bottle
18-605-185 4 x 2.5 L Glass Bottle
18-605-186 4 x 2.5 L Poly-Coated Glass Bottle
18-605-187 4 x 4 L Poly-Coated Glass Bottle
18-605-188 20 L Poly Drum
18-605-189 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-605-178 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PH175500MLGL
Add to Cart
Add to Cart

PH175, 7664-38-2, H3PO4

Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

CAS Max % 85 to 88%, 12 to 15%
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 85 to 88%
Grade NF
Chemical Name or Material Phosphoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.