Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 85 to 88%, 12 to 15% |
| Packaging | Poly-Coated Glass Bottle |
| Quantity | 4 x 4 L |
| Percent Purity | 85 to 88% |
| Chemical Name or Material | Phosphoric Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
